CAS RN 6601-22-5
IUPAC Name:
2,4-dichloro-6-(morpholin-4-yl)-1,3,5-triazine
Synonyms:
2,4-dichloro-6-(morpholin-4-yl)-1,3,5-triazine
Molecular Weight (g/mol):
235.07
Molecular Formula:
C7H8Cl2N4O
InChi Key:
UQAMDAUJTXFNAD-UHFFFAOYSA-N
SMILES:
ClC1=NC(=NC(Cl)=N1)N1CCOCC1
1
–
1
of
1
results
1
2,4-Dichloro-6-morpholino-1,3,5-triazine 98.0+%, TCI America™
CAS: 6601-22-5 Molecular Formula: C7H8Cl2N4O Molecular Weight (g/mol): 235.068 MDL Number: MFCD00460178 InChI Key: UQAMDAUJTXFNAD-UHFFFAOYSA-N Synonym: 2,4-dichloro-6-morpholino-1,3,5-triazine,4-4,6-dichloro-1,3,5-triazin-2-yl morpholine,unii-ndh2046uk7,2,4-dichloro-6-4-morpholinyl-1,3,5-triazine,2,4-dichloro-6-morpholin-4-yl-1,3,5-triazine,1,3,5-triazine, 2,4-dichloro-6-4-morpholinyl,2,4-dichloro-6-morpholin-4-yl-1,3,5 triazine,pubchem21087,2,4-dichlor-6-morpholin-4-yl-1,3,5-triazin,2,6-dichloro-4-morpholino-1,3,5-triazine PubChem CID: 81076 IUPAC Name: 4-(4,6-dichloro-1,3,5-triazin-2-yl)morpholine SMILES: C1COCCN1C2=NC(=NC(=N2)Cl)Cl