missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylaniline, 97%

CAS: 30273-00-8 | C7H7Cl2N | 176.04 g/mol

Supplier:  Thermo Scientific Chemicals A1259114

 View more versions of this product

Catalog No. AAA1259114


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichloro-6-methylaniline
43°C to 45°C
C7H7Cl2N
25 g
4,6-dichloro-2-methylaniline, benzenamine, 2,4-dichloro-6-methyl, acmc-20aodo, 4,6-dichloro-o-toluidine, 2-amino-3,5-dichlorotoluene, 2,4-dichloro-6-methylphenylamine, 4,6-dichloro-2-methylphenylamine, 2,4-bis chloranyl-6-methyl-aniline, 2,4-dichloro-6-methylaniline
CC1=CC(=CC(=C1N)Cl)Cl
176.04
176.05
30273-00-8
82°C to 84°C (0.1 mmHg)
MFCD00044074
2208221
UAISVUGQLKXPFF-UHFFFAOYSA-N
2,4-dichloro-6-methylaniline
1268287
97%

Chemical Identifiers

30273-00-8
176.04
UAISVUGQLKXPFF-UHFFFAOYSA-N
1268287
CC1=CC(=CC(=C1N)Cl)Cl
C7H7Cl2N
MFCD00044074
4,6-dichloro-2-methylaniline, benzenamine, 2,4-dichloro-6-methyl, acmc-20aodo, 4,6-dichloro-o-toluidine, 2-amino-3,5-dichlorotoluene, 2,4-dichloro-6-methylphenylamine, 4,6-dichloro-2-methylphenylamine, 2,4-bis chloranyl-6-methyl-aniline, 2,4-dichloro-6-methylaniline
2,4-dichloro-6-methylaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only