missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-iodoaniline, 97%

CAS: 697-90-5 | C6H4Cl2IN | 287.909 g/mol

$104.15 - $391.90

Chemical Identifiers

CAS 697-90-5
Molecular Formula C6H4Cl2IN
Molecular Weight (g/mol) 287.909
MDL Number MFCD00007661
InChI Key YJRSMJTVXWBFJJ-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-iodobenzenamine, 2,4-dichloro-6-iodophenylamine, 4,6-dichloro-2-iodophenylamine, 2,4-dichloro-6-iodo-phenylamine, benzenamine,2,4-dichloro-6-iodo, benzenamine, 2,4-dichloro-6-iodo, 1-amino-2,4-dichloro-6-iodobenzene
PubChem CID 7577525
IUPAC Name 2,4-dichloro-6-iodoaniline
SMILES C1=C(C=C(C(=C1Cl)N)I)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1765906
View Documents
Thermo Scientific Chemicals
L1765906
5 g
Each for $104.15
Only null left
 
AAL1765914
View Documents
Thermo Scientific Chemicals
L1765914
25 g
Each for $391.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

697-90-5
287.909
YJRSMJTVXWBFJJ-UHFFFAOYSA-N
7577525
C1=C(C=C(C(=C1Cl)N)I)Cl
C6H4Cl2IN
MFCD00007661
2,4-dichloro-6-iodobenzenamine, 2,4-dichloro-6-iodophenylamine, 4,6-dichloro-2-iodophenylamine, 2,4-dichloro-6-iodo-phenylamine, benzenamine,2,4-dichloro-6-iodo, benzenamine, 2,4-dichloro-6-iodo, 1-amino-2,4-dichloro-6-iodobenzene
2,4-dichloro-6-iodoaniline

Specifications

697-90-5
C6H4Cl2IN
5 g
3246130
2,4-dichloro-6-iodobenzenamine, 2,4-dichloro-6-iodophenylamine, 4,6-dichloro-2-iodophenylamine, 2,4-dichloro-6-iodo-phenylamine, benzenamine,2,4-dichloro-6-iodo, benzenamine, 2,4-dichloro-6-iodo, 1-amino-2,4-dichloro-6-iodobenzene
C1=C(C=C(C(=C1Cl)N)I)Cl
287.909
287.92
2,4-Dichloro-6-iodoaniline
79°C to 81°C
MFCD00007661
UN2811
Light sensitive
YJRSMJTVXWBFJJ-UHFFFAOYSA-N
2,4-dichloro-6-iodoaniline
7577525
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only