missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. D221325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D221325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D221325G Supplier TCI America Supplier No. D221325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 400-70-4
Molecular Formula C7H2Cl2F3NO2
Molecular Weight (g/mol) 259.993
MDL Number MFCD00007073
InChI Key VLVNHMVSVDVAOA-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-nitrobenzotrifluoride, 1,5-dichloro-2-nitro-4-trifluoromethyl benzene, 2,4-dichloro-3,5-nitrobenzotrifluoride, benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl, 2,4-dichloro-5-trifluoromethyl-1-nitrobenzene, pubchem4862, 2,4-dichloro-5-nitro benzotrifluoride, 3-trifluoromethyl-4,6-dichloronitrobenzene, 2,4-dichloro-5-trifluoromethyl nitrobenzene
PubChem CID 621179
IUPAC Name 1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F

Specifications

Melting Point 54°C
Color Yellow
Quantity 25 g
Formula Weight 259.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-5-nitrobenzotrifluoride