missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-5-methylpyrimidine 98.0+%, TCI America™
$42.87 - $283.33
Chemical Identifiers
| CAS | 1780-31-0 |
|---|---|
| Molecular Formula | C5H4Cl2N2 |
| Molecular Weight (g/mol) | 163.001 |
| MDL Number | MFCD00023197 |
| InChI Key | DQXNTSXKIUZJJS-UHFFFAOYSA-N |
| Synonym | pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p |
| PubChem CID | 74508 |
| IUPAC Name | 2,4-dichloro-5-methylpyrimidine |
| SMILES | CC1=CN=C(N=C1Cl)Cl |
Chemical Identifiers
| 1780-31-0 | |
| 163.001 | |
| DQXNTSXKIUZJJS-UHFFFAOYSA-N | |
| 74508 | |
| CC1=CN=C(N=C1Cl)Cl |
| C5H4Cl2N2 | |
| MFCD00023197 | |
| pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p | |
| 2,4-dichloro-5-methylpyrimidine |
Specifications
| 1780-31-0 | |
| 109°C | |
| MFCD00023197 | |
| pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p | |
| CC1=CN=C(N=C1Cl)Cl | |
| 163.001 | |
| 163.00 | |
| Crystalline Lumps |
| White-Yellow | |
| C5H4Cl2N2 | |
| 1 g | |
| DQXNTSXKIUZJJS-UHFFFAOYSA-N | |
| 2,4-dichloro-5-methylpyrimidine | |
| 74508 | |
| ≥98.0% (GC) | |
| 2,4-Dichloro-5-methylpyrimidine |
Safety and Handling
TSCA : Yes