accessibility menu, dialog, popup

UserName

CH3NClNCl

CAS RN 1780-31-0

IUPAC Name: 2,4-dichloro-5-methylpyrimidine
Synonyms: 2,4-dichloro-5-methylpyrimidine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: DQXNTSXKIUZJJS-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)N=C1Cl
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)