missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylpyrimidine 98.0+%, TCI America™

Catalog No. D36131G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36131G 1 g
D36135G 5 g
D361325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D36131G Supplier TCI America Supplier No. D36131G

Chemical Identifiers

CAS 1780-31-0
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00023197
InChI Key DQXNTSXKIUZJJS-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p
PubChem CID 74508
IUPAC Name 2,4-dichloro-5-methylpyrimidine
SMILES CC1=CN=C(N=C1Cl)Cl

Specifications

Color White-Yellow
Boiling Point 109°C
Quantity 1 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichloro-5-methylpyrimidine