missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylpyrimidine, 98%

Catalog No. AC398290050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398290050 5 g Glass bottle
AC398290010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398290050 Supplier Thermo Scientific Chemicals Supplier No. 398290050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1780-31-0 | C5H4Cl2N2 | 163.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1780-31-0
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.01
InChI Key DQXNTSXKIUZJJS-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p
PubChem CID 74508
IUPAC Name 2,4-dichloro-5-methylpyrimidine
SMILES CC1=CN=C(N=C1Cl)Cl

Specifications

Melting Point 26.0°C to 28.0°C
Density 1.3900g/mL
Boiling Point 109.0°C (11.0 mmHg)
Flash Point 113°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.39
Formula Weight 163.01
Percent Purity 98%
Chemical Name or Material 2, 4-Dichloro-5-methylpyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 217-227-8

RUO – Research Use Only