missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methoxyaniline, 98%

Catalog No. p-7028275
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2549703 1 g
AAB2549706 5 g
AAB2549714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2549703 Supplier Thermo Scientific Chemicals Supplier No. B2549703
Only null left

CAS: 98446-49-2 | C7H7Cl2NO | 192.039 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98446-49-2
Molecular Formula C7H7Cl2NO
Molecular Weight (g/mol) 192.039
MDL Number MFCD00974410
InChI Key AJROJTARXSATEB-UHFFFAOYSA-N
Synonym 5-amino-2,4-dichloroanisole, 2,4-dichloro-5-methoxybenzenamine, 4,6-dichloro-m-anisidine, 2,4-dichloro-5-methoxy-phenylamine, benzenamine, 2,4-dichloro-5-methoxy, 2,4-dichloro-5-methoxyphenylamine, dichloromethoxyaniline, pubchem4393, acmc-209sae, 4,5-dichloro-m-anisidine
PubChem CID 1476636
IUPAC Name 2,4-dichloro-5-methoxyaniline
SMILES COC1=C(C=C(C(=C1)N)Cl)Cl

Specifications

Quantity 1 g
UN Number UN2810
Formula Weight 192.05
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-methoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only