missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-5-fluorobenzoic Acid 98.0+%, TCI America™
$46.20 - $202.15
Chemical Identifiers
| CAS | 86522-89-6 |
|---|---|
| Molecular Formula | C7H2Cl2FO2 |
| Molecular Weight (g/mol) | 207.99 |
| MDL Number | MFCD00075281 |
| InChI Key | KZCWJHUTTSVCRO-UHFFFAOYSA-M |
| Synonym | 2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid |
| PubChem CID | 688138 |
| IUPAC Name | 2,4-dichloro-5-fluorobenzoate |
| SMILES | [O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl |
Chemical Identifiers
| 86522-89-6 | |
| 207.99 | |
| KZCWJHUTTSVCRO-UHFFFAOYSA-M | |
| 688138 | |
| [O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl |
| C7H2Cl2FO2 | |
| MFCD00075281 | |
| 2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid | |
| 2,4-dichloro-5-fluorobenzoate |
Specifications
| 86522-89-6 | |
| White-Yellow | |
| MFCD00075281 | |
| 2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid | |
| [O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl | |
| 207.99 | |
| 209.00 | |
| Crystalline Powder |
| 144°C | |
| C7H2Cl2FO2 | |
| 5 g | |
| KZCWJHUTTSVCRO-UHFFFAOYSA-M | |
| 2,4-dichloro-5-fluorobenzoate | |
| 688138 | |
| ≥98.0% (GC,T) | |
| 2,4-Dichloro-5-fluorobenzoic Acid |
Safety and Handling
TSCA : No