missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-3-ethyl-6-nitrophenol 98.0+%, TCI America™
Supplier: TCI America D351025G
Specifications
| 2,4-Dichloro-3-ethyl-6-nitrophenol | |
| 48°C | |
| MFCD00270764 | |
| 2811 | |
| YTVCECQSAPGJBB-UHFFFAOYSA-M | |
| 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate | |
| 7020339 | |
| ≥98.0% (GC,T) |
| 99817-36-4 | |
| C8H6Cl2NO3 | |
| 25 g | |
| 2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol | |
| CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O | |
| 235.04 | |
| 236.05 | |
| Crystalline Powder |
Chemical Identifiers
| 99817-36-4 | |
| 235.04 | |
| YTVCECQSAPGJBB-UHFFFAOYSA-M | |
| 7020339 | |
| CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O |
| C8H6Cl2NO3 | |
| MFCD00270764 | |
| 2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol | |
| 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate |
Safety and Handling
EINECSNumber : (3)-4208
TSCA : Yes