missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-naphthol 97.0+%, TCI America™

Catalog No. p-7126176
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D03835G Supplier TCI America Supplier No. D03835G

Chemical Identifiers

CAS 2050-76-2
Molecular Formula C10H6Cl2O
Molecular Weight (g/mol) 213.06
MDL Number MFCD00003931
InChI Key HVLJEMXDXOTWLV-UHFFFAOYSA-N
PubChem CID 16313
IUPAC Name 2,4-dichloronaphthalen-1-ol
SMILES OC1=C2C=CC=CC2=C(Cl)C=C1Cl

Specifications

Melting Point 105°C
Quantity 5 g
Synonym 2,4-dichloro-1-naphthol, 2,4-dichloronaphthol, 1-naphthalenol, 2,4-dichloro, 1-naphthalenol,2,4-dichloro, 2,4-dichloro-.alpha.-naphthol, 1-naphthol, 2,4-dichloro, aurora ka-5007, 1-naphthol,4-dichloro, acmc-1cdm5, 2,4-dichloronaphth-1-ol
Formula Weight 213.06
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-1-naphthol [for photography]
EINECSNumber (4)-0382
TSCA Yes