missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-iodobenzene 98.0+%, TCI America™

Catalog No. D144125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D144125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D144125G Supplier TCI America Supplier No. D144125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29898-32-6
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
MDL Number MFCD00001035
InChI Key ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-iodobenzene, 2,4-dichloroiodobenzene, benzene, 2,4-dichloro-1-iodo, 1-iodo-2,4-dichlorobenzene, pubchem3692, 2,4-dichlor-jodbenzol, acmc-209hbu, 1,3-dichloro4-iodobenzene, 1,3-dichloro 4-iodobenzene, 2,4-dichloro-1-iodo-benzene
PubChem CID 96864
IUPAC Name 2,4-dichloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)I

Specifications

Quantity 25 g
Formula Weight 272.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-1-iodobenzene