missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-iodobenzene, 97%, Thermo Scientific™

Catalog No. p-2696990
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00185EE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00185EE Supplier Thermo Scientific Chemicals Supplier No. TL00185EE

Chemical Identifiers

CAS 29898-32-6
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
InChI Key ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-iodobenzene, 2,4-dichloroiodobenzene, benzene, 2,4-dichloro-1-iodo, 1-iodo-2,4-dichlorobenzene, pubchem3692, 2,4-dichlor-jodbenzol, acmc-209hbu, 1,3-dichloro4-iodobenzene, 1,3-dichloro 4-iodobenzene, 2,4-dichloro-1-iodo-benzene
PubChem CID 96864
IUPAC Name 2,4-dichloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)I

Specifications

CAS Min % 97.0
Density 2.0100g/mL
Assay Percent Range 97%
Quantity 50 g
Formula Weight 272.9
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloro-1-iodobenzene
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.