missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-(4-nitrophenoxy)benzene, 98%

Catalog No. p-7024771
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1997714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1997714 Supplier Thermo Scientific Chemicals Supplier No. A1997714
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 1836-75-5 | C12H7Cl2NO3 | 284.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1836-75-5
Molecular Formula C12H7Cl2NO3
Molecular Weight (g/mol) 284.092
MDL Number MFCD00128026
InChI Key XITQUSLLOSKDTB-UHFFFAOYSA-N
Synonym nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
PubChem CID 15787
IUPAC Name 2,4-dichloro-1-(4-nitrophenoxy)benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl

Specifications

Melting Point 68°C to 71°C
Quantity 25 g
UN Number UN3077
Beilstein 1887356
Merck Index 14,6598
Formula Weight 284.1
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-1-(4-nitrophenoxy)benzene
Hazard Category H302-H350-H360D
Hazard Statement GHS H Statement
H350-H360-H302
May cause cancer.
May damage fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 217-406-0
RTECSNumber KN8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only