Learn More
2,4-Dibromophenol, 99%
CAS: 615-58-7 | C6H4Br2O | 251.91 g/mol
$56.71 - $56.71
Chemical Identifiers
| CAS | 615-58-7 |
|---|---|
| Molecular Formula | C6H4Br2O |
| Molecular Weight (g/mol) | 251.91 |
| MDL Number | MFCD00002149 |
| InChI Key | FAXWFCTVSHEODL-UHFFFAOYSA-N |
| Synonym | phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol |
| PubChem CID | 12005 |
| ChEBI | CHEBI:34238 |
| IUPAC Name | 2,4-dibromophenol |
| SMILES | C1=CC(=C(C=C1Br)Br)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC406510050
|
Thermo Scientific Chemicals
406510050 |
5 g | Glass bottle |
Each for $56.71
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 615-58-7 | |
| 251.91 | |
| FAXWFCTVSHEODL-UHFFFAOYSA-N | |
| 12005 | |
| 2,4-dibromophenol |
| C6H4Br2O | |
| MFCD00002149 | |
| phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol | |
| CHEBI:34238 | |
| C1=CC(=C(C=C1Br)Br)O |
Specifications
| 615-58-7 | |
| 100.0 | |
| Beige or White to Yellow | |
| >110°C | |
| 98.5% min. (GC) | |
| C6H4Br2O | |
| MFCD00002149 | |
| phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol | |
| C1=CC(=C(C=C1Br)Br)O | |
| 251.91 | |
| CHEBI:34238 | |
| 99% | |
| 2, 4-Dibromophenol, 99% |
| 98.5 | |
| 36°C to 39°C | |
| 154°C (11.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| Br2C6H3OH | |
| 5 g | |
| FAXWFCTVSHEODL-UHFFFAOYSA-N | |
| 2,4-dibromophenol | |
| 12005 | |
| 251.91 | |
| Crystalline Mass, Chunks or Crystals and Powder |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,i
GHS Signal Word: Danger
EINECSNumber : 210-436-5
RTECSNumber : SK8010000
TSCA : TSCA
RUO – Research Use Only