missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromobenzyl Alcohol 98.0+%, TCI America™

Catalog No. D50025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50025G 5 g
D50021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50025G Supplier TCI America Supplier No. D50025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 666747-06-4
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.93
MDL Number MFCD09996906
InChI Key VXEVXBMLHQPKGA-UHFFFAOYSA-N
Synonym 2,4-dibromophenyl methanol, 2,4-dibromobenzyl alcohol, benzenemethanol, 2,4-dibromo, 2,4-dibromobenzenemethanol, benzenemethanol,2,4-dibromo, pubchem9781, 2,4-dibromobenzylalcohol, acmc-1bfu5, 2,4-bis bromanyl phenyl methanol
PubChem CID 17836195
IUPAC Name (2,4-dibromophenyl)methanol
SMILES OCC1=C(Br)C=C(Br)C=C1

Specifications

Melting Point 82°C
Quantity 5 g
Formula Weight 265.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromobenzyl Alcohol