missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromobenzoic Acid 98.0+%, TCI America™

Catalog No. D42781G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D42781G 1 g
D42785G 5 g
2 options

Catalog No. D42781G

Supplier: TCI America D42781G

Only null left

Chemical Identifiers

CAS 611-00-7
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00234253
InChI Key NAGGYODWMPFKJQ-UHFFFAOYSA-N
PubChem CID 11896
IUPAC Name 2,4-dibromobenzoic acid
SMILES OC(=O)C1=CC=C(Br)C=C1Br

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 279.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromobenzoic Acid
EINECSNumber (3)-0062
TSCA No