missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromoaniline 98.0+%, TCI America™

Catalog No. D115310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D115310G 10 g
D115325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D115310G Supplier TCI America Supplier No. D115310G

Chemical Identifiers

CAS 615-57-6
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00007633
InChI Key DYSRXWYRUJCNFI-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dibromo, 2,4-dibromobenzenamine, 2,4-dibromo-phenylamine, dibromobenzenamine, 2,4-dibromanilin, pubchem3447, 2,4-dibromophenylamine, benzenamine,4-dibromo, 2,4-bis bromanyl aniline, acmc-209mu0
PubChem CID 12004
IUPAC Name 2,4-dibromoaniline
SMILES C1=CC(=C(C=C1Br)Br)N

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 156°C
Quantity 10 g
UN Number 2811
Formula Weight 250.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromoaniline