missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dibromoacetophenone, 98%

Catalog No. AC112680100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112680100 10 g Glass bottle
AC112680500 50 g Glass bottle
AC112682500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112680100 Supplier Thermo Scientific Chemicals Supplier No. 112680100

CAS: 99-73-0 | C8H6Br2O | 277.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-73-0
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.94
MDL Number MFCD00000200
InChI Key FKJSFKCZZIXQIP-UHFFFAOYSA-N
Synonym 2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
PubChem CID 7454
IUPAC Name 2-bromo-1-(4-bromophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 107°C to 111°C
Color Yellow to Beige
Boiling Point 1415°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH2Br
Quantity 10 g
Beilstein 07, 285
Fieser 01,77; 03,34
Merck Index 15, 1438
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in warm alcohol,soluble in ethyl alcohol,ethyl ether,chloroform
Formula Weight 277.94
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4'-Dibromoacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-783-6

RUO – Research Use Only