missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dibromoacetophenone, 98+%

CAS: 99-73-0 | C8H6Br2O | 277.943 g/mol

Supplier:  Thermo Scientific Chemicals A1368714

 View more versions of this product

Catalog No. AAA1368714


Only null left
Explore available promotions

Description

Description

2,4'-Dibromoacetophenone is used as a reagent for analysis of fatty acids by HPLC as 4-bromophenacyl esters, for use in the protection of phenols and carboxylic acids. It undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. It is also useful in the esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4′-Dibromoacetophenone is used as a reagent for analysis of fatty acids by HPLC as 4-bromophenacyl esters, for use in the protection of phenols and carboxylic acids. It undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. It is also useful in the esterification of carboxylic acids.

Solubility
Soluble in dimethyl sulfoxide (5 mg/ml), methanol (20 mg/ml), toluene and ethanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with bases and oxidizing agents.
Specifications

Specifications

2,4'-Dibromoacetophenone
108°C to 112°C
C8H6Br2O
25 g
607604
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
FKJSFKCZZIXQIP-UHFFFAOYSA-N
2-bromo-1-(4-bromophenyl)ethanone
7454
≥98%
99-73-0
Pungent
MFCD00000200
UN3261
14,1427
Soluble in dimethyl sulfoxide (5mg/ml),methanol (20mg/ml),toluene and ethanol. Insoluble in water.
C1=CC(=CC=C1C(=O)CBr)Br
277.943
277.95

Chemical Identifiers

99-73-0
277.943
FKJSFKCZZIXQIP-UHFFFAOYSA-N
7454
C1=CC(=CC=C1C(=O)CBr)Br
C8H6Br2O
MFCD00000200
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
2-bromo-1-(4-bromophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 202-783-6

RTECSNumber : AM6950000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only