missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-fluoroaniline, 97%, Thermo Scientific Chemicals

Catalog No. p-7021905
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1443514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1443514 Supplier Thermo Scientific Chemicals Supplier No. A1443514

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141474-37-5
Molecular Formula C6H4Br2FN
Molecular Weight (g/mol) 268.91
MDL Number MFCD00042230
InChI Key YJLXEKFYZIBUPJ-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dibromo-6-fluoro, 2,4-dibromo-6-fluorobenzenamine, 2,4-dibromo-6-fluoro-phenylamine, 2,4-dibromo-6-fluoro aniline, pubchem3442, intermediates-zcf02078, ksc494o1h, 2,4-dibromo-6-fluoro-aniline, acmc-1c814, 4,6-dibromo-2-fluorophenylamine
PubChem CID 2736765
IUPAC Name 2,4-dibromo-6-fluoroaniline
SMILES NC1=C(F)C=C(Br)C=C1Br

Specifications

Melting Point 62°C to 64°C
Quantity 25 g
UN Number UN2811
Beilstein 6289292
Formula Weight 268.92
Percent Purity 97%
Chemical Name or Material 2,4-Dibromo-6-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only