missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1-nitrobenzene 98.0+%, TCI America™

Catalog No. D47901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47901G 1 g
D47905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47901G Supplier TCI America Supplier No. D47901G
Only null left

Chemical Identifiers

CAS 51686-78-3
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.903
MDL Number MFCD02660416
InChI Key DXRVYZGVVFZCFP-UHFFFAOYSA-N
Synonym 2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene
PubChem CID 12315646
IUPAC Name 2,4-dibromo-1-nitrobenzene
SMILES C1=CC(=C(C=C1Br)Br)[N+](=O)[O-]

Specifications

Melting Point 61°C
Color Yellow
Quantity 1 g
Formula Weight 280.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromo-1-nitrobenzene