accessibility menu, dialog, popup

UserName

BrBrNOO

CAS RN 51686-78-3

IUPAC Name: 2,4-dibromo-1-nitrobenzene
Synonyms: 2,4-dibromo-1-nitrobenzene
Molecular Weight (g/mol): 280.9
Molecular Formula: C6H3Br2NO2
InChi Key: DXRVYZGVVFZCFP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Br)C=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)