missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1,3,5-trimethylbenzene 98.0+%, TCI America™

Catalog No. p-7128713
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52625G 5 g
D526225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52625G Supplier TCI America Supplier No. D52625G

Chemical Identifiers

CAS 6942-99-0
Molecular Formula C9H10Br2
Molecular Weight (g/mol) 277.987
MDL Number MFCD00013525
InChI Key CIHJFEWFZJQTFE-UHFFFAOYSA-N
Synonym 2,4-dibromomesitylene, benzene, 2,4-dibromo-1,3,5-trimethyl, mesitylene, 2,4-dibromo, p-brommesitylbromid, mesitylene,4-dibromo, acmc-1bag9, ksc494c7h, benzene,4-dibromo-1,3,5-trimethyl, #
PubChem CID 81368
IUPAC Name 2,4-dibromo-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1Br)C)Br)C

Specifications

Melting Point 63°C
Color White
Boiling Point 279°C
Quantity 5 g
Formula Weight 277.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromo-1,3,5-trimethylbenzene