missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibromo-1,3,5-trimethylbenzene 98.0+%, TCI America™
Supplier: TCI America D526225G
Specifications
| 2,4-Dibromo-1,3,5-trimethylbenzene | |
| 63°C | |
| 279°C | |
| MFCD00013525 | |
| 2,4-dibromomesitylene, benzene, 2,4-dibromo-1,3,5-trimethyl, mesitylene, 2,4-dibromo, p-brommesitylbromid, mesitylene,4-dibromo, acmc-1bag9, ksc494c7h, benzene,4-dibromo-1,3,5-trimethyl, # | |
| CC1=CC(=C(C(=C1Br)C)Br)C | |
| 277.987 | |
| 277.99 | |
| Crystalline Powder |
| 6942-99-0 | |
| White | |
| C9H10Br2 | |
| 25 g | |
| CIHJFEWFZJQTFE-UHFFFAOYSA-N | |
| 2,4-dibromo-1,3,5-trimethylbenzene | |
| 81368 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6942-99-0 | |
| 277.987 | |
| CIHJFEWFZJQTFE-UHFFFAOYSA-N | |
| 81368 | |
| CC1=CC(=C(C(=C1Br)C)Br)C |
| C9H10Br2 | |
| MFCD00013525 | |
| 2,4-dibromomesitylene, benzene, 2,4-dibromo-1,3,5-trimethyl, mesitylene, 2,4-dibromo, p-brommesitylbromid, mesitylene,4-dibromo, acmc-1bag9, ksc494c7h, benzene,4-dibromo-1,3,5-trimethyl, # | |
| 2,4-dibromo-1,3,5-trimethylbenzene |
Safety and Handling
TSCA : No