missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine, 97%

Catalog No. AAB2574203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2574203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2574203 Supplier Thermo Scientific Chemicals Supplier No. B2574203

CAS: 43167-79-9 | C19H20N6O | 348.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 43167-79-9
Molecular Formula C19H20N6O
Molecular Weight (g/mol) 348.41
MDL Number MFCD00272362
InChI Key GSXOUQFSYTZEJF-UHFFFAOYSA-N
Synonym 1,3,5-triazine-2,4-diamine, 6-4-morpholinyl-n2,n4-diphenyl, 2,4-dianilino-6-4-morpholinyl-1,3,5-triazine, 6-morpholino-n2,n4-diphenyl-1,3,5-triazine-2,4-diamine, 6-morpholin-4-yl-n,n'-diphenyl-1,3,5 triazine-2,4-diamine, 6-morpholin-4-yl-n2,n4-diphenyl-1,3,5-triazine-2,4-diamine, aamt, 2,4-bis anilino-6-4-morpholinyl-1,3,5-triazine, 6-4-morpholinyl-n2,n4-diphenyl-1,3,5-triazine-2,4-diamine
PubChem CID 780750
IUPAC Name 6-morpholin-4-yl-2-N,4-N-diphenyl-1,3,5-triazine-2,4-diamine
SMILES C1COCCN1C2=NC(=NC(=N2)NC3=CC=CC=C3)NC4=CC=CC=C4

Specifications

Melting Point 194°C to 196°C
Quantity 1 g
Formula Weight 348.41
Percent Purity 97%
Chemical Name or Material 2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only