accessibility menu, dialog, popup

UserName

ONNNNHNHN

CAS RN 43167-79-9

IUPAC Name: 6-(morpholin-4-yl)-N2,N4-diphenyl-1,3,5-triazine-2,4-diamine
Synonyms: 6-(morpholin-4-yl)-N2,N4-diphenyl-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 348.41
Molecular Formula: C19H20N6O
InChi Key: GSXOUQFSYTZEJF-UHFFFAOYSA-N
SMILES: C1CN(CCO1)C1=NC(NC2=CC=CC=C2)=NC(NC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results