missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Diaminopyrimidine 98.0+%, TCI America™
Supplier: TCI America D212025G
Specifications
| 2,4-Diaminopyrimidine | |
| 149°C | |
| C4H6N4 | |
| 25 g | |
| YAAWASYJIRZXSZ-UHFFFAOYSA-N | |
| pyrimidine-2,4-diamine | |
| 67431 | |
| 110.12 | |
| Crystalline Powder |
| 156-81-0 | |
| White-Yellow | |
| MFCD00038023 | |
| 2,4-diaminopyrimidine, 2,4-pyrimidinediamine, 2,6-diaminopyrimidine, pyrimidinediamine, diaminopyrimidine, pyrimidine, 2,4-diamino, 2,4-diamino-pyrimidine, pyrimidine-2,4-diyldiamine, 2,4-pyrimidinediamine 9ci, 4-amino-2-pyrimidineamine | |
| C1=CN=C(N=C1N)N | |
| 110.12 | |
| CHEBI:43745 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 156-81-0 | |
| 110.12 | |
| YAAWASYJIRZXSZ-UHFFFAOYSA-N | |
| 67431 | |
| pyrimidine-2,4-diamine |
| C4H6N4 | |
| MFCD00038023 | |
| 2,4-diaminopyrimidine, 2,4-pyrimidinediamine, 2,6-diaminopyrimidine, pyrimidinediamine, diaminopyrimidine, pyrimidine, 2,4-diamino, 2,4-diamino-pyrimidine, pyrimidine-2,4-diyldiamine, 2,4-pyrimidinediamine 9ci, 4-amino-2-pyrimidineamine | |
| CHEBI:43745 | |
| C1=CN=C(N=C1N)N |
Safety and Handling
TSCA : No