CAS RN 156-81-0
CAS RN 156-81-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,4-Diaminopyrimidine 98.0+%, TCI America™
CAS: 156-81-0 Molecular Formula: C4H6N4 Molecular Weight (g/mol): 110.12 MDL Number: MFCD00038023 InChI Key: YAAWASYJIRZXSZ-UHFFFAOYSA-N Synonym: 2,4-diaminopyrimidine,2,4-pyrimidinediamine,2,6-diaminopyrimidine,pyrimidinediamine,diaminopyrimidine,pyrimidine, 2,4-diamino,2,4-diamino-pyrimidine,pyrimidine-2,4-diyldiamine,2,4-pyrimidinediamine 9ci,4-amino-2-pyrimidineamine PubChem CID: 67431 ChEBI: CHEBI:43745 IUPAC Name: pyrimidine-2,4-diamine SMILES: C1=CN=C(N=C1N)N