missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminopyrimidine 98.0+%, TCI America™

Catalog No. D21205G
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D21205G 5 g
D212025G 25 g
2 options

Catalog No. D21205G

Supplier: TCI America D21205G

Only null left

Chemical Identifiers

CAS 156-81-0
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD00038023
InChI Key YAAWASYJIRZXSZ-UHFFFAOYSA-N
Synonym 2,4-diaminopyrimidine, 2,4-pyrimidinediamine, 2,6-diaminopyrimidine, pyrimidinediamine, diaminopyrimidine, pyrimidine, 2,4-diamino, 2,4-diamino-pyrimidine, pyrimidine-2,4-diyldiamine, 2,4-pyrimidinediamine 9ci, 4-amino-2-pyrimidineamine
PubChem CID 67431
ChEBI CHEBI:43745
IUPAC Name pyrimidine-2,4-diamine
SMILES C1=CN=C(N=C1N)N

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 5 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diaminopyrimidine