missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminophenol Sulfate 98.0+%, TCI America™

Catalog No. d249925g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D249925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D249925G Supplier TCI America Supplier No. D249925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74283-34-4
Molecular Formula C6H10N2O5S
Molecular Weight (g/mol) 222.215
MDL Number MFCD01321156
InChI Key JKMWKYDJCPSJSI-UHFFFAOYSA-N
PubChem CID 17922877
IUPAC Name 2,4-diaminophenol;sulfuric acid
SMILES C1=CC(=C(C=C1N)N)O.OS(=O)(=O)O

Specifications

Color Black-Gray
Quantity 25 g
Formula Weight 222.22
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diaminophenol Sulfate
EINECSNumber (3)-2772
TSCA No