missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminoanisole, MilliporeSigma™ Supelco™

Catalog No. 111019714
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019714 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019714 Supplier Merck Emd Millipore Supplier No. 32831100MG

2,4-Diaminoanisole belongs to the class of primary aromatic amines (PAAs). It is categorized as a carcinogenic compound.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 615-05-4
Physical Form Neat
Linear Formula CH3 OC6H3(NH2)2
MDL Number MFCD00007803
Synonym 4-Methoxy-m-phenylenediamine
Type Analytical Standard
Quantity 100 mg
Formula Weight 138.17
Grade Analytical Standard