missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-methoxy-1,3,5-triazine, TCI America™

Catalog No. D22325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22325G Supplier TCI America Supplier No. D22325G
Only null left

Chemical Identifiers

CAS 2827-45-4
Molecular Formula C4H7N5O
Molecular Weight (g/mol) 141.13
MDL Number MFCD00191340
InChI Key XVMFICQRQHBOOT-UHFFFAOYSA-N
PubChem CID 17803
ChEBI CHEBI:38930
IUPAC Name 6-methoxy-1,3,5-triazine-2,4-diamine
SMILES COC1=NC(N)=NC(N)=N1

Specifications

Color White
Quantity 5 g
Formula Weight 141.13
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-methoxy-1,3,5-triazine
RTECSNumber XY6890000
TSCA No