accessibility menu, dialog, popup

UserName

CAS RN 2827-45-4

H3CONNH2NH2NN

CAS RN 2827-45-4

IUPAC Name: 6-methoxy-1,3,5-triazine-2,4-diamine
Synonyms: 6-methoxy-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 141.13
Molecular Formula: C4H7N5O
InChi Key: XVMFICQRQHBOOT-UHFFFAOYSA-N
SMILES: COC1=NC(N)=NC(N)=N1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results