missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-isopropoxy-1,3,5-triazine, TCI America™

Catalog No. D22251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22251G Supplier TCI America Supplier No. D22251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24860-40-0
Molecular Formula C6H11N5O
Molecular Weight (g/mol) 169.19
MDL Number MFCD00191339
InChI Key ATCQNYLEZRQALQ-UHFFFAOYSA-N
PubChem CID 32722
IUPAC Name 6-(propan-2-yloxy)-1,3,5-triazine-2,4-diamine
SMILES CC(C)OC1=NC(N)=NC(N)=N1

Specifications

Melting Point 172°C
Color White
Quantity 1 g
Formula Weight 169.19
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-isopropoxy-1,3,5-triazine
RTECSNumber XY6880000
TSCA Yes