accessibility menu, dialog, popup

UserName

CH3H3CONNH2NH2NN

CAS RN 24860-40-0

IUPAC Name: 6-(propan-2-yloxy)-1,3,5-triazine-2,4-diamine
Synonyms: 6-isopropoxy-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 169.19
Molecular Formula: C6H11N5O
InChi Key: ATCQNYLEZRQALQ-UHFFFAOYSA-N
SMILES: CC(C)OC1=NC(N)=NC(N)=N1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results