missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-hydroxypyrimidine 98.0+%, TCI America™

Catalog No. D132125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D132125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D132125G Supplier TCI America Supplier No. D132125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-06-4
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.119
MDL Number MFCD00006098
InChI Key SWELIMKTDYHAOY-UHFFFAOYSA-N
Synonym 2,4-diamino-6-hydroxypyrimidine, 2,6-diaminopyrimidin-4-ol, 2,6-diaminopyrimidin-4 1h-one, 2,6-diamino-4-pyrimidinol, 2,6-diamino-4-hydroxypyrimidine, 2,6-diaminopyrimidin-4 3h-one, 2,6-diamino-4-pyrimidinone, 4 1h-pyrimidinone, 2,6-diamino, 2,4-diaminohypoxanthine, 6-aminoisocytosine
PubChem CID 2944
IUPAC Name 2,6-diamino-1H-pyrimidin-4-one
SMILES C1=C(NC(=NC1=O)N)N

Specifications

Melting Point 286°C
Quantity 25 g
Formula Weight 126.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-hydroxypyrimidine