accessibility menu, dialog, popup

UserName

CAS RN 56-06-4

IUPAC Name: 2,6-diamino-1,4-dihydropyrimidin-4-one
Synonyms: 6-aminoisocytosine
Molecular Weight (g/mol): 126.12
Molecular Formula: C4H6N4O
InChi Key: SWELIMKTDYHAOY-UHFFFAOYSA-N
SMILES: NC1=CC(=O)N=C(N)N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)