missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-hydroxypyrimidine, 96%, Thermo Scientific™

Chemical Identifiers

CAS 56-06-4
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006098
InChI Key SWELIMKTDYHAOY-UHFFFAOYSA-N
Synonym 2,4-diamino-6-hydroxypyrimidine, 2,6-diaminopyrimidin-4-ol, 2,6-diaminopyrimidin-4 1h-one, 2,6-diamino-4-pyrimidinol, 2,6-diamino-4-hydroxypyrimidine, 2,6-diaminopyrimidin-4 3h-one, 2,6-diamino-4-pyrimidinone, 4 1h-pyrimidinone, 2,6-diamino, 2,4-diaminohypoxanthine, 6-aminoisocytosine
PubChem CID 2944
IUPAC Name 2,6-diamino-1H-pyrimidin-4-one
SMILES C1=C(NC(=NC1=O)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC112240250
SDS View Documents
Thermo Scientific Chemicals
112240250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC112241000
SDS View Documents
Thermo Scientific Chemicals
112241000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

56-06-4
126.12
SWELIMKTDYHAOY-UHFFFAOYSA-N
2944
C1=C(NC(=NC1=O)N)N
C4H6N4O
MFCD00006098
2,4-diamino-6-hydroxypyrimidine, 2,6-diaminopyrimidin-4-ol, 2,6-diaminopyrimidin-4 1h-one, 2,6-diamino-4-pyrimidinol, 2,6-diamino-4-hydroxypyrimidine, 2,6-diaminopyrimidin-4 3h-one, 2,6-diamino-4-pyrimidinone, 4 1h-pyrimidinone, 2,6-diamino, 2,4-diaminohypoxanthine, 6-aminoisocytosine
2,6-diamino-1H-pyrimidin-4-one

Specifications

56-06-4
100.0
96%
C4H6N4O
25g
15, 2983
Solubility in water: 11.5g/L (20°C).
C1=C(NC(=NC1=O)N)N
126.12
126.12
2,4-Diamino-6-hydroxypyrimidine, 96%
95.0
Authentic
Plastic bottle
MFCD00006098
24, 469
2,4-diamino-6-hydroxypyrimidine, 2,6-diaminopyrimidin-4-ol, 2,6-diaminopyrimidin-4 1h-one, 2,6-diamino-4-pyrimidinol, 2,6-diamino-4-hydroxypyrimidine, 2,6-diaminopyrimidin-4 3h-one, 2,6-diamino-4-pyrimidinone, 4 1h-pyrimidinone, 2,6-diamino, 2,4-diaminohypoxanthine, 6-aminoisocytosine
SWELIMKTDYHAOY-UHFFFAOYSA-N
2,6-diamino-1H-pyrimidin-4-one
2944
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-254-4

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only