missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-hydroxypyrimidine, 96%

Catalog No. AC112240250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112240250 25 g
AC112241000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112240250 Supplier Thermo Scientific Chemicals Supplier No. 112240250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56-06-4 | C4H6N4O | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-06-4
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006098
InChI Key SWELIMKTDYHAOY-UHFFFAOYSA-N
Synonym 2,4-diamino-6-hydroxypyrimidine, 2,6-diaminopyrimidin-4-ol, 2,6-diaminopyrimidin-4 1h-one, 2,6-diamino-4-pyrimidinol, 2,6-diamino-4-hydroxypyrimidine, 2,6-diaminopyrimidin-4 3h-one, 2,6-diamino-4-pyrimidinone, 4 1h-pyrimidinone, 2,6-diamino, 2,4-diaminohypoxanthine, 6-aminoisocytosine
PubChem CID 2944
IUPAC Name 2,6-diamino-1H-pyrimidin-4-one
SMILES C1=C(NC(=NC1=O)N)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 270.0°C to 290.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Quantity 25 g
Beilstein 24, 469
Merck Index 15, 2983
Solubility Information Solubility in water: 11.5g/L (20°C).
Formula Weight 126.12
Percent Purity 96%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-hydroxypyrimidine, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-254-4
TSCA TSCA

RUO – Research Use Only