missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di-tert-butylphenol 97.0+%, TCI America™

Catalog No. D022925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D022925G 25 g
D0229500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D022925G Supplier TCI America Supplier No. D022925G

Chemical Identifiers

CAS 96-76-4
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.329
MDL Number MFCD00008828
InChI Key ICKWICRCANNIBI-UHFFFAOYSA-N
Synonym 2,4-di-tert-butylphenol, 2,4-di-t-butylphenol, phenol, 2,4-bis 1,1-dimethylethyl, antioxidant no. 33, 1-hydroxy-2,4-di-tert-butylbenzene, phenol, 2,4-di-tert-butyl, 2,4-bis tert-butyl phenol, prodox 146, prodox 146a-85x, unii-fob94g6hzt
PubChem CID 7311
IUPAC Name 2,4-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)C(C)(C)C

Specifications

Melting Point 56°C
Boiling Point 261°C
Quantity 25 g
UN Number 3077
Formula Weight 206.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Di-tert-butylphenol
EINECSNumber (3)-0521&(3)-0526
RTECSNumber SK8260000
TSCA Yes