missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di-tert-butylphenol, 97%

CAS: 96-76-4 | C14H22O | 206.329 g/mol

Supplier:  Thermo Scientific Chemicals A185090B

 View more versions of this product

Catalog No. AAA185090B


Only null left
Explore available promotions

Description

Description

2,4-Di-tert-butylphenol is used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts. It is a raw material for phosphite anti-oxidants. The antioxidants are primarily used in the manufacture of polyolefins. It is also used as an intermediate for the preparation of UV stabilizers and in manufacturing of pharmaceuticals and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Di-tert-butylphenol is used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts. It is a raw material for phosphite anti-oxidants. The antioxidants are primarily used in the manufacture of polyolefins. It is also used as an intermediate for the preparation of UV stabilizers and in manufacturing of pharmaceuticals and fragrances.

Solubility
Soluble in methanol, ether, acetone, chloroform. 2,4-Di-tert-butylphenol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2,4-Di-tert-butylphenol
53°C to 57°C
265°C
C14H22O
1000 g
1910383
Soluble in methanol,ether,acetone,chloroform. 2,4-Di-tert-butylphenol. Insoluble in water.
CC(C)(C)C1=CC(=C(C=C1)O)C(C)(C)C
206.329
206.33
96-76-4
0.887
129°C (264°F)
MFCD00008828
UN3077
2,4-di-tert-butylphenol, 2,4-di-t-butylphenol, phenol, 2,4-bis 1,1-dimethylethyl, antioxidant no. 33, 1-hydroxy-2,4-di-tert-butylbenzene, phenol, 2,4-di-tert-butyl, 2,4-bis tert-butyl phenol, prodox 146, prodox 146a-85x, unii-fob94g6hzt
ICKWICRCANNIBI-UHFFFAOYSA-N
2,4-ditert-butylphenol
7311
97%

Chemical Identifiers

96-76-4
206.329
ICKWICRCANNIBI-UHFFFAOYSA-N
7311
CC(C)(C)C1=CC(=C(C=C1)O)C(C)(C)C
C14H22O
MFCD00008828
2,4-di-tert-butylphenol, 2,4-di-t-butylphenol, phenol, 2,4-bis 1,1-dimethylethyl, antioxidant no. 33, 1-hydroxy-2,4-di-tert-butylbenzene, phenol, 2,4-di-tert-butyl, 2,4-bis tert-butyl phenol, prodox 146, prodox 146a-85x, unii-fob94g6hzt
2,4-ditert-butylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c

H302-H315-H317-H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-532-0

RTECSNumber : SK8260000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only