missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Di-tert-amylphenol 98.0+%, TCI America™
$46.20 - $150.16
Chemical Identifiers
| CAS | 120-95-6 |
|---|---|
| Molecular Formula | C16H26O |
| Molecular Weight (g/mol) | 234.38 |
| MDL Number | MFCD00041929 |
| InChI Key | WMVJWKURWRGJCI-UHFFFAOYSA-N |
| Synonym | 2,4-di-tert-pentylphenol, 2,4-di-tert-amylphenol, 2,4-di-t-pentylphenol, 2,4-bis 1,1-dimethylpropyl phenol, phenol, 2,4-bis 1,1-dimethylpropyl, di-tert-amylphenol, prodox 156, phenol, 2,4-di-tert-pentyl, 2,4-bis 2-methylbutan-2-yl phenol, unii-852f1hp88h |
| PubChem CID | 8455 |
| IUPAC Name | 2,4-bis(2-methylbutan-2-yl)phenol |
| SMILES | CCC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)CC |
Chemical Identifiers
| 120-95-6 | |
| 234.38 | |
| WMVJWKURWRGJCI-UHFFFAOYSA-N | |
| 8455 | |
| CCC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)CC |
| C16H26O | |
| MFCD00041929 | |
| 2,4-di-tert-pentylphenol, 2,4-di-tert-amylphenol, 2,4-di-t-pentylphenol, 2,4-bis 1,1-dimethylpropyl phenol, phenol, 2,4-bis 1,1-dimethylpropyl, di-tert-amylphenol, prodox 156, phenol, 2,4-di-tert-pentyl, 2,4-bis 2-methylbutan-2-yl phenol, unii-852f1hp88h | |
| 2,4-bis(2-methylbutan-2-yl)phenol |
Specifications
| 120-95-6 | |
| 143°C | |
| MFCD00041929 | |
| 3077 | |
| WMVJWKURWRGJCI-UHFFFAOYSA-N | |
| 2,4-bis(2-methylbutan-2-yl)phenol | |
| 8455 | |
| ≥98.0% (GC) | |
| 2,4-Di-tert-amylphenol |
| White | |
| C16H26O | |
| 5 g | |
| 2,4-di-tert-pentylphenol, 2,4-di-tert-amylphenol, 2,4-di-t-pentylphenol, 2,4-bis 1,1-dimethylpropyl phenol, phenol, 2,4-bis 1,1-dimethylpropyl, di-tert-amylphenol, prodox 156, phenol, 2,4-di-tert-pentyl, 2,4-bis 2-methylbutan-2-yl phenol, unii-852f1hp88h | |
| CCC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)CC | |
| 234.38 | |
| 234.38 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-0521&(3)-0526
RTECSNumber : SL3500000
TSCA : Yes