missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane

Catalog No. p-91306
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC337470050 5 g Glass bottle
AC337470250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC337470050 Supplier Thermo Scientific Chemicals Supplier No. 337470050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126519-89-9 | C11H17ClO5SSi | 324.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126519-89-9 , 75-09-2
Molecular Formula C11H17ClO5SSi
Molecular Weight (g/mol) 324.85
MDL Number MFCD00054774
InChI Key NYIDSUMRGUILGR-UHFFFAOYSA-N
Synonym 2-4-chlorosulfonylphenyl ethyltrimethoxysilane, 4-2-trimethoxysilylethyl benzenesulfonyl chloride, 4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride, benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl, 2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid, 4-2-trimethoxysilyl ethyl benzenesulfonyl chloride, acmc-20ete8, 4-chlorosulfonyl phenethyltrimethoxysilane
PubChem CID 2733807
IUPAC Name 4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
SMILES CO[Si](CCC1=CC=C(C=C1)S(Cl)(=O)=O)(OC)OC

Specifications

CAS Min % 45.0
CAS Max % 55.0
Color Yellow
Boiling Point >40.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: reacts
Formula Weight 324.85
Percent Purity 45 to 55% (solids)
Physical Form Solution
Chemical Name or Material 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 279-267-2
TSCA TSCA

RUO – Research Use Only