missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenylthio)benzaldehyde, 98%

Catalog No. p-7020782
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAA1285206 5 g
AAA1285214 25 g
2 options

Catalog No. AAA1285206

Supplier: Thermo Scientific Chemicals A1285206

Only null left

CAS: 107572-07-6 | C13H9ClOS | 248.724 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107572-07-6
Molecular Formula C13H9ClOS
Molecular Weight (g/mol) 248.724
MDL Number MFCD00051766
InChI Key CQKLAEUCMKGSEQ-UHFFFAOYSA-N
Synonym 2-4-chlorophenylthio benzaldehyde, 2-4-chlorophenyl thio benzaldehyde, 2-4-chlorophenyl sulfanyl benzaldehyde, 2-4-chlorophenyl sulfanylbenzaldehyde, benzaldehyde,2-4-chlorophenyl thio, acmc-20aopf, 2-4-chlorophenyl sulphanyl benzaldehyde
PubChem CID 2778624
IUPAC Name 2-(4-chlorophenyl)sulfanylbenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)SC2=CC=C(C=C2)Cl

Specifications

Melting Point 71°C to 74°C
Boiling Point 102°C to 104°C (14 mmHg)
Quantity 5 g
Beilstein 5810711
Sensitivity Air Sensitive
Formula Weight 248.72
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenylthio)benzaldehyde
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only