missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenylthio)benzaldehyde, 98%

Catalog No. AAA1285206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1285206 5 g
AAA1285214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1285206 Supplier Thermo Scientific Chemicals Supplier No. A1285206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107572-07-6 | C13H9ClOS | 248.724 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107572-07-6
Molecular Formula C13H9ClOS
Molecular Weight (g/mol) 248.724
MDL Number MFCD00051766
InChI Key CQKLAEUCMKGSEQ-UHFFFAOYSA-N
Synonym 2-4-chlorophenylthio benzaldehyde, 2-4-chlorophenyl thio benzaldehyde, 2-4-chlorophenyl sulfanyl benzaldehyde, 2-4-chlorophenyl sulfanylbenzaldehyde, benzaldehyde,2-4-chlorophenyl thio, acmc-20aopf, 2-4-chlorophenyl sulphanyl benzaldehyde
PubChem CID 2778624
IUPAC Name 2-(4-chlorophenyl)sulfanylbenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)SC2=CC=C(C=C2)Cl

Specifications

Melting Point 71°C to 74°C
Boiling Point 102°C to 104°C (14 mmHg)
Quantity 5 g
Beilstein 5810711
Sensitivity Air Sensitive
Formula Weight 248.72
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenylthio)benzaldehyde
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only