missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Chlorophenyl)benzimidazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 303°C |
| Quantity | 5 g |
| Formula Weight | 228.68 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(4-Chlorophenyl)benzimidazole |
Safety and Handling
| TSCA | No |