accessibility menu, dialog, popup

UserName

HNNCl

CAS RN 1019-85-8

IUPAC Name: 2-(4-chlorophenyl)-1H-1,3-benzodiazole
Synonyms: 2-(4-chlorophenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 228.68
Molecular Formula: C13H9ClN2
InChi Key: PTXUJRTVWRYYTE-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results