missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)benzimidazole 98.0+%, TCI America™

Catalog No. C20271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20271G 1 g
C20275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20271G Supplier TCI America Supplier No. C20271G
Only null left

Chemical Identifiers

CAS 1019-85-8
Molecular Formula C13H9ClN2
Molecular Weight (g/mol) 228.679
InChI Key PTXUJRTVWRYYTE-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl benzimidazole, 2-4-chlorophenyl-1h-benzoimidazole, 2-4-chlorophenyl-1h-benzimidazole, 2-4-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-chlorophenyl, 2-4-chlorophenyl-1h-benzo d imidazole, 2-4-chlorophenyl-1h-1,3-benzodiazole, acmc-2097xz, 1h-benzimidazole,2-4-chlorophenyl
PubChem CID 759417
IUPAC Name 2-(4-chlorophenyl)-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)Cl

Specifications

Melting Point 303°C
Quantity 1 g
Formula Weight 228.68
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chlorophenyl)benzimidazole